Learn More
Hach Company SC4x00 UV Protection Screen
Catalog No. 5000207164 Shop All Hach Company Products
Encompass
SC4x00 UV Protection ScreenIncludes This assembly covers the face of the SC4200c/SC4500 protecting it from the sun The transparent door can be opened for operating the controller It is UV absorbent and protects the controller from harmful UV rays
Catalog No. 50-002-07164 Supplier Hach Company Supplier No. LXZ524.99.00004
SC4x00 UV Protection ScreenIncludes This assembly covers the face of the SC4200c/SC4500 protecting it from the sun The transparent door can be opened for operating the controller It is UV absorbent and protects the controller from harmful UV rays