Learn More
Selleck Chemical LLC SC66 10mg 871361-88-5
SDP
Supplier:  Selleck Chemical LLC S531310MG
Encompass_Preferred
SC66 is an allosteric inhibitor which displays a dual-inhibitory function toward AKT activity with IC50 values of 0.77, 2.85 and 0.47 ?g/ml in HepG2, Huh7 and Hep3B cells after 72 h treatment, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8390