Learn More
Medchemexpress LLC HY-19832 5mg Medchemexpress, SC66 CAS:871361-88-5 Purity:>98%
SDP
Catalog No. 502082192
Encompass_Preferred
Medchemexpress, HY-19832 5mg SC66 CAS:871361-88-5 SC66 is an Akt inhibitor, reduces cell viability in a dose- and time-dependent manner, inhibits colony formation and induces apoptosis in hepatocellular carcinoma (HCC) cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2192 Supplier Medchemexpress LLC Supplier No. HY198325MG
Medchemexpress, HY-19832 5mg SC66 CAS:871361-88-5 SC66 is an Akt inhibitor, reduces cell viability in a dose- and time-dependent manner, inhibits colony formation and induces apoptosis in hepatocellular carcinoma (HCC) cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.