Learn More
Medchemexpress LLC HY-18749 10mg , SC79 CAS:305834-79-1 Purity:>98%
SDP
Catalog No. 502259205
Encompass_Preferred
HY-18749 10mg SC79 CAS: 305834-79-1 SC79, a unique specific and BBB permeable Akt activator, activates Akt in the cytosol and inhibits Akt membrane translocation. SC79 specifically binds to the PH domain of Akt. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9205 Supplier Medchemexpress LLC Supplier No. HY1874910MG
HY-18749 10mg SC79 CAS: 305834-79-1 SC79, a unique specific and BBB permeable Akt activator, activates Akt in the cytosol and inhibits Akt membrane translocation. SC79 specifically binds to the PH domain of Akt. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.