Learn More
Selleck Chemical LLC SC79 10mg 305834-79-1
SDP
Supplier:  Selleck Chemical LLC S786310MG
Encompass_Preferred
SC79 is a brain-penetrable Akt phosphorylation activator and an inhibitor of Akt-PH domain translocation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5349