Learn More
Selleck Chemical LLC SC79
SDP
Catalog No. 5000002934
Encompass_Preferred
SC79 is a brain-penetrable Akt phosphorylation activator and an inhibitor of Akt-PH domain translocation
Catalog No. 50-000-02934 Supplier Selleck Chemical LLC Supplier No. S7863200MG
SC79 is a brain-penetrable Akt phosphorylation activator and an inhibitor of Akt-PH domain translocation