Learn More
TARGETMOL CHEMICALS INC SC79 25MG
SDP
Catalog No. 502580929
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SC79 a selective Akt activator may be used to enhance Akt activity in various pathological and physiological conditions. purity: 98%
Catalog No. 50-258-0929 Supplier TARGETMOL CHEMICALS INC Supplier No. T227425MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SC79 a selective Akt activator may be used to enhance Akt activity in various pathological and physiological conditions. purity: 98%