Learn More
Radiometer America Radiometer ABL90 FLEX PLUS Sensor Cassette with Automatic Quality Management (AQM) - 600 Tests, Full Panel CO-OX
SDP
Catalog No. NC1169803
Encompass
ABL90 FLEX PLUS Sensor Cassette (600 tests)
600 tests, 30 day Sensor Cassette for the ABL90 FLEX PLUS blood gas analyzer
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.
Catalog No. NC1169803 Supplier Radiometer America Supplier No. 946008
ABL90 FLEX PLUS Sensor Cassette (600 tests)
600 tests, 30 day Sensor Cassette for the ABL90 FLEX PLUS blood gas analyzer
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.