Learn More
Salimetrics LLC SCA SALIVARY COLL AID 50/PK
Catalog No. NC0751801
Encompass
Saliva Collection Aid, 50/PK Size: 55 mm x 12 mm (approx.) The Saliva Collection Aid (SCA) is a new device that fits securely inside common externally threaded cryovials to simplify collection of whole saliva samples from participants. Developed by SalivaBio, LLC in collaboration with the Center for Interdisciplinary Salivary Bioscience Research at the Johns Hopkins University (Press Release), the new collection aid is now available from Salimetrics. *Simple and easy to use *Increased participant compliance *Individually packaged in a clean, foil pouch *One-time use and dispose *Ready-to-go instructions *Vented design helps avoid sample foaming *Constructed of polypropylene to avoid sample retention or contamination *Universal fit with common external thread cryovials *Allows sample collection directly into cryovials in the field
Catalog No. NC0751801 Supplier Salimetrics LLC Supplier No. 5016.04
Saliva Collection Aid, 50/PK Size: 55 mm x 12 mm (approx.) The Saliva Collection Aid (SCA) is a new device that fits securely inside common externally threaded cryovials to simplify collection of whole saliva samples from participants. Developed by SalivaBio, LLC in collaboration with the Center for Interdisciplinary Salivary Bioscience Research at the Johns Hopkins University (Press Release), the new collection aid is now available from Salimetrics. *Simple and easy to use *Increased participant compliance *Individually packaged in a clean, foil pouch *One-time use and dispose *Ready-to-go instructions *Vented design helps avoid sample foaming *Constructed of polypropylene to avoid sample retention or contamination *Universal fit with common external thread cryovials *Allows sample collection directly into cryovials in the field