Learn More
Aviva Systems Biology Corp Scaf8 Antibody - C-terminal region (ARP40791_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40791_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Scaf8. It was validated on Western Blot. . Alternate names: A630086M08Rik, A930036P18Rik, AA408306, AI447644, AI448652, AU015035, KIAA1116, mKIAA1116, Rbm16ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-284-892