Learn More
Grainger Adam Equipment Carrying Case, Hard Shell, Die Cut Foam, 1 ea
Catalog No. 502795280
Encompass_Preferred
This robust hard carrying case is designed to protect Triple Beam Balances during transport. Featuring a durable hard shell construction, the case includes a locking mechanism to keep the instrument secure. The interior is equipped with die cut foam inserts that provide designated storage for the balance, weights, and tripod legs.
  • Durable hard shell construction
  • Includes lock for security
  • Features die cut foam inserts for organized storage of balance, weights, and tripod legs
  • Compatible with TBB 2610S, TBB 2610T, and TBB 610S models
  • Constructed from aluminum, plastic, and foam materials
Catalog No. 50-279-5280 Supplier Grainger Supplier No. 19YP01
This robust hard carrying case is designed to protect Triple Beam Balances during transport. Featuring a durable hard shell construction, the case includes a locking mechanism to keep the instrument secure. The interior is equipped with die cut foam inserts that provide designated storage for the balance, weights, and tripod legs.
  • Durable hard shell construction
  • Includes lock for security
  • Features die cut foam inserts for organized storage of balance, weights, and tripod legs
  • Compatible with TBB 2610S, TBB 2610T, and TBB 610S models
  • Constructed from aluminum, plastic, and foam materials