Learn More
METTLER TOLEDO 3P INDUSTRIAL Protective Cover MS-TS
Catalog No. NC1119614
Encompass_Preferred
Protects Against Dust and Dirt: The protective cover decreases the need to clean your balance and helps prevents cross contamination. Simply replace as required.Safer Weighing Processes: The chemically resistant cover protects the balance against toxic or hazardous substances and provides operators with an increased level of safety.Easy to Install and Remove
Catalog No. NC1119614 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 30216667
Protects Against Dust and Dirt: The protective cover decreases the need to clean your balance and helps prevents cross contamination. Simply replace as required.Safer Weighing Processes: The chemically resistant cover protects the balance against toxic or hazardous substances and provides operators with an increased level of safety.Easy to Install and Remove