Learn More
Scientific Industries SCALE, PILL COUNTER & KIT
Catalog No. 501139344
Encompass_Preferred
KIT combines DRX-4C scale with RXP-4 thermal printer and calibration weight to optimize pill counting producing a receipt which includes transaction details such as, date, time, NDC number, and actual count. Capacity 320g x 0.001g. Class II pharmacy balance, NTEP certified as legal for trade in prescription weighing.
Catalog No. 50-113-9344 Supplier Scientific Industries Supplier No. DRX4C320 KIT
KIT combines DRX-4C scale with RXP-4 thermal printer and calibration weight to optimize pill counting producing a receipt which includes transaction details such as, date, time, NDC number, and actual count. Capacity 320g x 0.001g. Class II pharmacy balance, NTEP certified as legal for trade in prescription weighing.