Learn More
Scientific Industries Torbal AG2000 Laboratory Balance, 2000G x 0.01G
Catalog No. 502120109
Encompass_Preferred
2000g x 0.01g w/ Automatic Internal Calibration, ideal for weighing small loads, Auto calibration keeps the scale calibrated at all times. The scale includes 12 weighing functions, equipped with USB, RS232, and PS2 interface ports
This item is not returnable. View return policy
Catalog No. 502120109 Supplier Scientific Industries Supplier No. AG2000
2000g x 0.01g w/ Automatic Internal Calibration, ideal for weighing small loads, Auto calibration keeps the scale calibrated at all times. The scale includes 12 weighing functions, equipped with USB, RS232, and PS2 interface ports