Learn More
Electron Microscopy Sciences Scale Reticle RET 10, 21 mm, 10 mm Scale, 0.1 mm x 100 Divisions
SDP
Catalog No. 501901749
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 10 mm
Divisions: 0.1 mm x 100
Catalog No. 50-190-1749 Supplier Electron Microscopy Sciences Supplier No. 6027321
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 10 mm
Divisions: 0.1 mm x 100