Learn More
Electron Microscopy Sciences Scale Reticle RET 10, 19 mm, 10 mm Scale, 0.1 mm x 100 Divisions
SDP
Catalog No. 501901748
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 10 mm
Divisions: 0.1 mm x 100
Catalog No. 50-190-1748 Supplier Electron Microscopy Sciences Supplier No. 6027319
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 10 mm
Divisions: 0.1 mm x 100