Learn More
Electron Microscopy Sciences Scale Reticle RET 11, 21 mm, 5 mm Scale, 0.05 mm x 100 Divisions
SDP
Catalog No. 501902078
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 5 mm
Divisions: 0.05 mm x 100
Catalog No. 50-190-2078 Supplier Electron Microscopy Sciences Supplier No. 6027221
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 5 mm
Divisions: 0.05 mm x 100