Learn More
Electron Microscopy Sciences Scale Reticle RET 12, 19 mm, 20 mm Scale, 0.10 mm x 200 Divisions
SDP
Catalog No. 501901752
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 20 mm
Divisions: 0.10 mm x 200
Catalog No. 50-190-1752 Supplier Electron Microscopy Sciences Supplier No. 6027619
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 20 mm
Divisions: 0.10 mm x 200