Learn More
Electron Microscopy Sciences Scale Reticle RET 14, 21 mm, 2 mm Scale, 0.01 mm x 200 Divisions
SDP
Catalog No. 501901558
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 2 mm
Divisions: 0.01 mm x 200
Catalog No. 50-190-1558 Supplier Electron Microscopy Sciences Supplier No. 6027021
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 2 mm
Divisions: 0.01 mm x 200