Learn More
Electron Microscopy Sciences Scale Reticle RET 15, 19 mm, 5 mm Scale, 0.1 mm x 50 Divisions
SDP
Catalog No. 501901559
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 5 mm
Divisions: 0.1 mm x 50
Catalog No. 50-190-1559 Supplier Electron Microscopy Sciences Supplier No. 6027119
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Scale: 5 mm
Divisions: 0.1 mm x 50