Learn More
FUJIFILM Biosciences SCALEVIEW A2 500ML
Catalog No. 5000450369
Encompass_Preferred
SCALEVIEW-A2 is reagent used for tissue clearing by ScaleA2 method And the reagent used for antibody staining AbScale method and fluorescent staining ChemScale method of transparent tissue combining with SCALEVIEW-S series Their methods were developed by Dr Atsushi Miyawaki and others RIKEN Fixed mouse and rat brain is made transparent only soaking into the reagent Also the reagent used for immersion liquid and to enable microscopic observation 2mm section can be made transparent in 2 weeks using the reagents
Catalog No. 50-004-50369 Supplier FUJIFILM Biosciences Supplier No. 19318455
SCALEVIEW-A2 is reagent used for tissue clearing by ScaleA2 method And the reagent used for antibody staining AbScale method and fluorescent staining ChemScale method of transparent tissue combining with SCALEVIEW-S series Their methods were developed by Dr Atsushi Miyawaki and others RIKEN Fixed mouse and rat brain is made transparent only soaking into the reagent Also the reagent used for immersion liquid and to enable microscopic observation 2mm section can be made transparent in 2 weeks using the reagents