Learn More
FUJIFILM Biosciences SCALEVIEW S Trial Kit
Catalog No. 5000450427
Encompass_Preferred
SCALEVIEW-S Trial Kit consists of tissue clearing reagents used for ScaleS a tissue clearing method developed by Dr Atsushi Miyawaki and his colleagues This method uses solutions with mainly urea The tissue is made transparent and adjusted RI for microscopic observation by soaking in 6 kinds of solutions SCALEVIEW-S0 SCALEVIEW-S1 SCALEVIEW-S2 SCALEVIEW-S3 SCALEVIEW-S4 SCALEVIEW-SMt 2mm section can be made transparent in 1 day using the reagents The method can make a transparent tissue keeping the structure of proteins Therefore it is possible to observe the tissue expressing fluorescent proteins and to label by antibody and/or fluorescent dye after tissue clearing Its also applicable to not only mouse and rat brain but also human after dye The transparent tissue can be observed for electric microscope or confocal microscope
Catalog No. 50-004-50427 Supplier FUJIFILM Biosciences Supplier No. 29979901
SCALEVIEW-S Trial Kit consists of tissue clearing reagents used for ScaleS a tissue clearing method developed by Dr Atsushi Miyawaki and his colleagues This method uses solutions with mainly urea The tissue is made transparent and adjusted RI for microscopic observation by soaking in 6 kinds of solutions SCALEVIEW-S0 SCALEVIEW-S1 SCALEVIEW-S2 SCALEVIEW-S3 SCALEVIEW-S4 SCALEVIEW-SMt 2mm section can be made transparent in 1 day using the reagents The method can make a transparent tissue keeping the structure of proteins Therefore it is possible to observe the tissue expressing fluorescent proteins and to label by antibody and/or fluorescent dye after tissue clearing Its also applicable to not only mouse and rat brain but also human after dye The transparent tissue can be observed for electric microscope or confocal microscope