Learn More
World Precision Instrument Standard Scalpel Blade #10
SDP
Catalog No. 50822418
Encompass
Surgical blades are made of hardened and tempered steel or carbon steel. This kind of material is crucial in order to achieve a the high degree of sharpness required for a perfect first tissue incision. Other choices for surgical blades include titanium, ceramic, diamond and obsidian. Each box contains 100 sterile and stainless steel blades, and they are individually wrapped.
Catalog No. 50-822-418 Supplier World Precision Instrument Supplier No. 500239
Surgical blades are made of hardened and tempered steel or carbon steel. This kind of material is crucial in order to achieve a the high degree of sharpness required for a perfect first tissue incision. Other choices for surgical blades include titanium, ceramic, diamond and obsidian. Each box contains 100 sterile and stainless steel blades, and they are individually wrapped.