Learn More
Electron Microscopy Sciences Scalpel Disposable Blade Remover
SDP
Catalog No. 50949107
Encompass_Preferred
This device is designed for safe and easy removal of used disposable blades from any size or type of scalpel handle. It improves safety and hygiene by the elimination of direct hand-blade contact. Depending on the size, it can store 150-300 used blades.
Catalog No. 50-949-107 Supplier Electron Microscopy Sciences Supplier No. 72070
This device is designed for safe and easy removal of used disposable blades from any size or type of scalpel handle. It improves safety and hygiene by the elimination of direct hand-blade contact. Depending on the size, it can store 150-300 used blades.