Learn More
Fine Science Tools Scalpel Blade Remover, Stainless Steel
SDP
Catalog No. NC9590375
Encompass
Product: Scalpel Blade Remover/ Alloy / Material: Stainless Steel. Used to safely remove contaminated scalpel blades. Can be used for scalpels that fit the #3, #4, #7 and gross anatomy handles.
Catalog No. NC9590375 Supplier Fine Science Tools Supplier No. 1000200
Product: Scalpel Blade Remover/ Alloy / Material: Stainless Steel. Used to safely remove contaminated scalpel blades. Can be used for scalpels that fit the #3, #4, #7 and gross anatomy handles.