Learn More
Fine Science Tools Scalpel Blades - #11, 100/pk
SDP
Catalog No. NC9021088
Encompass
Product: Scalpel Blades - #11/ Blade Shape: Angled/ Cutting Edge: 20mm/ Thickness: 0.4mm/ Alloy / Material: Carbon Steel. Blades come individually wrapped in transparent packages. The alloy has a high carbon content which provides a longer lasting cutting edge. Not autoclavable. 100/pk
Catalog No. NC9021088 Supplier Fine Science Tools Supplier No. 1001100
Product: Scalpel Blades - #11/ Blade Shape: Angled/ Cutting Edge: 20mm/ Thickness: 0.4mm/ Alloy / Material: Carbon Steel. Blades come individually wrapped in transparent packages. The alloy has a high carbon content which provides a longer lasting cutting edge. Not autoclavable. 100/pk