Learn More
Surgical Design™ Scalpel Blades, Size 10 R - Box of 100, Stainless Steel
Catalog No. 21125081 Shop All Surgical Design Products
Change view
Click to view available options
Blade Material:
Stainless Steel
Catalog No. Blade Material
21-125-081 Stainless Steel
1 options
Catalog No. 21-125-081 Supplier Surgical Design™ Supplier No. SBSS10RBX
Add to Cart
Add to Cart
Size 10R stainless-steel scalpel blades (box of 100) are individually sterile-packed for detailed, controlled incisions with reliable sharpness.
  • Size 10R Specialized rounded-tip profile for controlled, shallow incisions and fine tissue work.
  • Stainless Steel Construction Resists corrosion and maintains a sharp, consistent cutting edge.
  • Sterile and Individually Wrapped Ensures safety, cleanliness, and easy single-use handling.
  • Standard Fitment Compatible with most No. 3 scalpel handles.
  • Box of 100 Convenient bulk packaging for clinics, labs, and classrooms.
  • Medical-Grade Quality Suitable for surgical, clinical, veterinary, and educational applications.
Specifications
Blade Material Stainless Steel
Product Type Scalpel Blade
For Use With (Equipment) No.3 Handle
Disposable Reusable
Sterility Sterile
Certifications/Compliance ISO/GMP
Autoclavable Autoclavable
Graduated No