Learn More
Med Vet International Miltex Disposable Safety Scalpel Stainless Steel, Retractable Blade, #10, 10/Box, 4-510
Catalog No. 502830947
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Disposable Safety Scalpels With Stainless Steel Retractable Blade. A scalpel is a bladed surgical instrument used to make cuts into the body. This is a very sharp instrument and comes in various sizes for different types of cuts and surgeries. Specifications:#10, 10/Box
Catalog No. 50-283-0947 Supplier Med Vet International Supplier No. MIL4510
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Disposable Safety Scalpels With Stainless Steel Retractable Blade. A scalpel is a bladed surgical instrument used to make cuts into the body. This is a very sharp instrument and comes in various sizes for different types of cuts and surgeries. Specifications:#10, 10/Box