Learn More
Med Vet International Dynarex Medicut Scalpels Disposable Sterile, #15, 10/Box
Catalog No. 502830724
Encompass_Preferred
Dynarex Medicut Scalpels feature a stainless steel blade with a finely honed cutting edge. The sterile scalpels are individually foil packed for convenience. Dynarex Medicut Scalpels are designed for improved sharpness and exacting performance.
Catalog No. 50-283-0724 Supplier Med Vet International Supplier No. 4115
Dynarex Medicut Scalpels feature a stainless steel blade with a finely honed cutting edge. The sterile scalpels are individually foil packed for convenience. Dynarex Medicut Scalpels are designed for improved sharpness and exacting performance.