Learn More
Med Vet International Exel Disposable Scalpel #10, stainless steel, 10/Box
Catalog No. 502643627
Encompass_Preferred
These products do not contain Natural Rubber Latex.Computer controlled grinding, sharpening and polishing, ensures consistently smooth incision propertiesSterilized by GAMMA radiation 2.5 – RADSA Vapor Corrosion Inhibitor (VCI) paper lining, leakage testing, and an aluminum foil peal pack help insure quality and integrity when ready for useEach production lot is sampled and conforms to ISO 2859, inspection level (S4) AQL 1.0 & 2.5Blade rigidity is tested to eliminate blade breakageBlades conform to ISO: 7740 (1995) & BS: 2982 (1986)Blades are ultrasonically cleaned prior to packingMade from premium quality Swiss carbon steel and British Stainless Steel
This item is not returnable. View return policy
Catalog No. 50-264-3627 Supplier Med Vet International Supplier No. 29550
These products do not contain Natural Rubber Latex.Computer controlled grinding, sharpening and polishing, ensures consistently smooth incision propertiesSterilized by GAMMA radiation 2.5 – RADSA Vapor Corrosion Inhibitor (VCI) paper lining, leakage testing, and an aluminum foil peal pack help insure quality and integrity when ready for useEach production lot is sampled and conforms to ISO 2859, inspection level (S4) AQL 1.0 & 2.5Blade rigidity is tested to eliminate blade breakageBlades conform to ISO: 7740 (1995) & BS: 2982 (1986)Blades are ultrasonically cleaned prior to packingMade from premium quality Swiss carbon steel and British Stainless Steel