Learn More
Sklar Instruments Scalpel Handle, 4-3/4 in., #3, Serrated, Stainless Steel, EA.Sklar Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection.
SDP
Catalog No. 501109706 Shop All Sklar Instruments Products
Encompass_Preferred
Scalpel Handle, 4-3/4 in., #3, Serrated, Stainless Steel, EA.Sklar's Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations for thumb and forefinger gripping. There are no metric markings on this particular #3 handle.
Catalog No. 50-110-9706 Supplier Sklar Instruments Supplier No. 062903
Scalpel Handle, 4-3/4 in., #3, Serrated, Stainless Steel, EA.Sklar's Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations for thumb and forefinger gripping. There are no metric markings on this particular #3 handle.