Learn More
Med Vet International Miltex #3 Scalpel Handle, Fits Blades 10-15
Catalog No. 502522295
Encompass_Preferred
The Miltex 4-7 #3 scalpel handle is made of stainless steel and has mm and cm graduations. This knife handle fits blades from size 10 to 15.
Catalog No. 50-252-2295 Supplier Med Vet International Supplier No. MIL47
The Miltex 4-7 #3 scalpel handle is made of stainless steel and has mm and cm graduations. This knife handle fits blades from size 10 to 15.