Learn More
DR Instruments Scalpel Handle #3 with No. 10 Blades
SDPSCI_ED
Catalog No. S72156
Change view
Click to view available options
Size:
# 10
Catalog No. Size
S72156 # 10
1 options
Catalog No. S72156 Supplier DR Instruments Supplier No. 27WB10BO
Add to Cart
Add to Cart
Scalpel Handle #3 comes with a 10 pack of No. 10 Blades.
  • Scalpel handle features black oxide coating for extra corrosion and rust resistance.
  • Includes number 10 blades, 10 pack.
  • Length: 5 in.
  • Ideal for dissection.
  • Ideal for wood carving.
Specifications
Blade Material Stainless Steel
Product Type Surgical Scalpel
For Use With (Application) Dissection, Wood Carving
Size # 10
Length (English) 5 in.
Length (Metric) 12.7 cm
Sterility Sterile
Includes Handle (# 3), Blades (# 10)
Handle Material Black Oxide Coated
Quantity 10 Pk.