Learn More
Sklar Instruments Scalpel Handle, Sklar 3K. Sklar #3K Scalpel Handle is a round, knurled, beaver style handle used for holding microsurgical series 50, 60, and 70 scalpel blades, EA.
SDP
Catalog No. 501094407 Shop All Sklar Instruments Products
Encompass_Preferred
Scalpel Handle, Sklar, #3K. Sklar's #3K Scalpel Handle is a round, knurled, beaver style handle used for holding microsurgical series 50, 60, and 70 scalpel blades. The tip of the handle is slotted for blade placement and screws in to hold the blade in place. It is often used in plastic surgery, podiatry, and other microsurgical procedures. This product is straight and 4-1/2 inches long. EA.
Catalog No. 50-109-4407 Supplier Sklar Instruments Supplier No. 971450
Scalpel Handle, Sklar, #3K. Sklar's #3K Scalpel Handle is a round, knurled, beaver style handle used for holding microsurgical series 50, 60, and 70 scalpel blades. The tip of the handle is slotted for blade placement and screws in to hold the blade in place. It is often used in plastic surgery, podiatry, and other microsurgical procedures. This product is straight and 4-1/2 inches long. EA.