Learn More
Mopec™ Scalpel Handle, #3L
SDP
Catalog No. 22445312 Shop All Mopec Products
Change view
Click to view available options
Size:
3L
Catalog No. Size
22-445-312 3L
1 options
Catalog No. 22-445-312 Supplier Mopec™ Supplier No. AJ142
Add to Cart
Add to Cart
Stainless steel scalpel handle for #10 thru #15 blades.
  • Durable and reusable
Specifications
Product Type Scalpel Handle
Size 3L
Disposable Reusable
Sterility Sterile
Graduated No