Learn More
Sklar Instruments Scalpel Handle,5-3/4 in., #4, Serrated, Stainless Steel, EA.Sklar #4 Scalpel Handle is similar to the #3 scalpel handle, but compatible with the larger size 20 series disposable blades. It is used for sharp dissection.
SDP
Catalog No. 501109707 Shop All Sklar Instruments Products
Encompass_Preferred
Scalpel Handle,5-3/4 in., #4, Serrated, Stainless Steel, EA.Sklar's #4 Scalpel Handle is similar to the #3 scalpel handle, but compatible with the larger size 20 series disposable blades. It is used for sharp dissection. There are centimeter and millimeter markings on the handle for measuring tissue and a center groove for thumb and forefinger gripping.
Catalog No. 50-110-9707 Supplier Sklar Instruments Supplier No. 062904
Scalpel Handle,5-3/4 in., #4, Serrated, Stainless Steel, EA.Sklar's #4 Scalpel Handle is similar to the #3 scalpel handle, but compatible with the larger size 20 series disposable blades. It is used for sharp dissection. There are centimeter and millimeter markings on the handle for measuring tissue and a center groove for thumb and forefinger gripping.