Learn More
Mopec Scalpel Handle, Bard Parker #8 
SDP
Catalog No. 22444268 Shop All Mopec Products
Change view
Click to view available options
Size:
5.5 in. (140 mm)
Catalog No. Size
22-444-268 5.5 in. (140 mm)
1 options
Catalog No. 22-444-268 Supplier Mopec Supplier No. AJ115
Add to Cart
Edge
Add to Cart
Smooth scalpel handle by Mopec for #60 and #70 blades. Durable and reusable.
  • Smooth
  • Scalpel Handle
  • Durable and reusable
Specifications
Blade Material Latex Free
Product Type Scalpel Handle
Size 5.5 in. (140 mm)
For Use With (Equipment) # 60 and # 70 Blades
Description Smooth