Learn More
Mopec Scalpel Handle for Double Edged Blade
SDP
Catalog No. 22444271 Shop All Mopec Products
Catalog No. 22-444-271 Supplier Mopec Supplier No. AJ127
Add to Cart
Add to Cart
Mopec's double edge blade handle is ideal for trimming surgical specimens and is a versatile tool for any laboratory. The double edge blade is securely held in place, yet easy to replace. The weighted pencil-style handle provides a firm, mobile grip.
  • Compatible with Mopec AJ111 double edge blades
  • 5 complimentary blades included
Specifications
Size AJ111 blades
For Use With (Equipment) Compatible with Mopec AJ111 Double Edge Blade