Learn More
Roboz Surgical SCALPEL HANDLE NO3 4IN
SDP
Catalog No. NC9202114
Encompass
This solid scalpel handle is designed to securely hold surgical blades for precise cutting tasks in laboratory environments. It is engineered for durability and consistent performance in research applications.
  • Features a standard number 3 configuration for compatibility with corresponding blades
  • Constructed as a solid, single-piece unit for enhanced stability
  • Measures 4 in. in total length for balanced handling
  • Designed specifically for animal research applications only
Catalog No. NC9202114 Supplier Roboz Surgical Supplier No. RS9843
This solid scalpel handle is designed to securely hold surgical blades for precise cutting tasks in laboratory environments. It is engineered for durability and consistent performance in research applications.
  • Features a standard number 3 configuration for compatibility with corresponding blades
  • Constructed as a solid, single-piece unit for enhanced stability
  • Measures 4 in. in total length for balanced handling
  • Designed specifically for animal research applications only