Learn More
Sklar Instruments Disposable Carbon Steel Surgical Blades, Sklar Sharp, Curved, #15C, BX100
SDP
Catalog No. 501181212 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Carbon Steel Surgical Blades, Sklar. Sharp, Curved, #15C. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #15C is most commonly used in dentistry and in periodontal procedures. The #15C blade features a longer cutting edge than the #15 blade and when combined with a long style handle provides an even more extended reach for the user. Disposable blades are available in bulk quantities of 100 per box.
Catalog No. 50-118-1212 Supplier Sklar Instruments Supplier No. 063016
Disposable Carbon Steel Surgical Blades, Sklar. Sharp, Curved, #15C. Sklar's Disposable, Sterile, Carbon Steel Surgical Blade #15C is most commonly used in dentistry and in periodontal procedures. The #15C blade features a longer cutting edge than the #15 blade and when combined with a long style handle provides an even more extended reach for the user. Disposable blades are available in bulk quantities of 100 per box.