Learn More
Sklar Instruments Scalpel Handle, Sklar 5 in., #3 with Metric Scale, Serrated, Stainless Steel.
SDP
Catalog No. 501181202 Shop All Sklar Instruments Products
Encompass_Preferred
Scalpel Handle, Sklar. 5 in., #3 with Metric Scale, Serrated, Stainless Steel. Sklar's Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations for thumb and forefinger gripping. This #3 handle has centimeter and millimeter markings on the handle for measuring tissue.
Catalog No. 50-118-1202 Supplier Sklar Instruments Supplier No. 062910
Scalpel Handle, Sklar. 5 in., #3 with Metric Scale, Serrated, Stainless Steel. Sklar's Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations for thumb and forefinger gripping. This #3 handle has centimeter and millimeter markings on the handle for measuring tissue.