Learn More
Sklar Instruments Scalpel Handle, Sklar 8-1/2 in., #3L, Serrated, Stainless Steel. Sklar Scalpel Handle #3L is longer and thinner than handle #3. EA.
SDP
Catalog No. 501181206 Shop All Sklar Instruments Products
Encompass_Preferred
Scalpel Handle, Sklar. 8-1/2 in., #3L, Serrated, Stainless Steel. Sklar's Scalpel Handle #3L is longer and thinner than handle #3. It is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations near the blade end and diagonal serrations at the opposite end for thumb and forefinger gripping.
Catalog No. 50-118-1206 Supplier Sklar Instruments Supplier No. 062915
Scalpel Handle, Sklar. 8-1/2 in., #3L, Serrated, Stainless Steel. Sklar's Scalpel Handle #3L is longer and thinner than handle #3. It is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations near the blade end and diagonal serrations at the opposite end for thumb and forefinger gripping.