Learn More
Sklar Instruments Scalpel Handle, Sklar 5-1/2 in., Stainless Steel. EA,
SDP
Catalog No. 501181201 Shop All Sklar Instruments Products
Encompass_Preferred
Scalpel Handle, Sklar. 5-1/2 in., Stainless Steel. Sklar's Angled Round Scalpel Handle is an angled handle that can be used with scalpel blades #10, #11, #12, and #15.
Catalog No. 50-118-1201 Supplier Sklar Instruments Supplier No. 062909
Scalpel Handle, Sklar. 5-1/2 in., Stainless Steel. Sklar's Angled Round Scalpel Handle is an angled handle that can be used with scalpel blades #10, #11, #12, and #15.