Learn More
Sklar Instruments Scalpel Handle, Sklar 10 in., Stainless Steel. Sklar Scalpel Handle #3XL is longer and thinner than handle #3L. EA.
SDP
Catalog No. 501181207 Shop All Sklar Instruments Products
Encompass_Preferred
Scalpel Handle, Sklar. 10 in., Stainless Steel. Sklar's Scalpel Handle #3XL is longer and thinner than handle #3L. It is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations near the blade end and diagonal serrations at the opposite end for thumb and forefinger gripping.
Catalog No. 50-118-1207 Supplier Sklar Instruments Supplier No. 062916
Scalpel Handle, Sklar. 10 in., Stainless Steel. Sklar's Scalpel Handle #3XL is longer and thinner than handle #3L. It is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations near the blade end and diagonal serrations at the opposite end for thumb and forefinger gripping.