Learn More
Sklar Instruments Scalpel Handle, Sklar 5-3/4 in., Stainless Steel. Scalpel Handle #8 to be used with Size 20 Series Blade. EA.
SDP
Catalog No. 501181204 Shop All Sklar Instruments Products
Encompass_Preferred
Scalpel Handle, Sklar. 5-3/4 in., Stainless Steel.Scalpel Handle #8 to be used with Size 20 Series Blade. Sklar's Scalpel Handle #8 has a slightly wider handle and is used with the size 20 series disposable blade.
Catalog No. 50-118-1204 Supplier Sklar Instruments Supplier No. 062912
Scalpel Handle, Sklar. 5-3/4 in., Stainless Steel.Scalpel Handle #8 to be used with Size 20 Series Blade. Sklar's Scalpel Handle #8 has a slightly wider handle and is used with the size 20 series disposable blade.