Learn More
Med Vet International Miltex #7 Scalpel Handle, Fits Blades 10-15
Catalog No. 502831786
Encompass_Preferred
The Miltex 4-9 #7 scalpel handle is made of stainless steel and has mm and cm graduations. This knife handle fits blades from size 10 to 15.
Catalog No. 50-283-1786 Supplier Med Vet International Supplier No. MIL49
The Miltex 4-9 #7 scalpel handle is made of stainless steel and has mm and cm graduations. This knife handle fits blades from size 10 to 15.