Learn More
World Precision Instrument Scalpel Handle #3, 13cm
SDP
Catalog No. 50822412
Encompass
Scalpel Handles are tagged with numbers to  denote the size and type of the blades that are compatible with the handle. The Scalpel Handle #3 is usually used with smaller blades that have a curved cutting edge, like the #10 or #15 replaceable surgical blades. It can also be with the pointed #11 blade. These handles and blade work well for microdissection or fine surgeries when making precise incisions. The Scalpel Handle #4 is a little larger and heavier than the #3. It gives you more control and the ability to apply more force. The #4 scalpel handle is often used with larger blades like the #20, #21, #22, #23, #24, and #25 blades. These blades have a similar shapes as the blades used with the #3 handle, but the blades are larger in size. A Scalpel Handle #7 is a smaller handle designed for precision and fine detail work. It is commonly used in ophthalmology, microdissection, and delicate procedures.
Catalog No. 50-822-412 Supplier World Precision Instrument Supplier No. 500236
Scalpel Handles are tagged with numbers to  denote the size and type of the blades that are compatible with the handle. The Scalpel Handle #3 is usually used with smaller blades that have a curved cutting edge, like the #10 or #15 replaceable surgical blades. It can also be with the pointed #11 blade. These handles and blade work well for microdissection or fine surgeries when making precise incisions. The Scalpel Handle #4 is a little larger and heavier than the #3. It gives you more control and the ability to apply more force. The #4 scalpel handle is often used with larger blades like the #20, #21, #22, #23, #24, and #25 blades. These blades have a similar shapes as the blades used with the #3 handle, but the blades are larger in size. A Scalpel Handle #7 is a smaller handle designed for precision and fine detail work. It is commonly used in ophthalmology, microdissection, and delicate procedures.