Learn More
Fine Science Tools Scalpel Handle - #4
SDP
Catalog No. NC9079870
Encompass
Product: Scalpel Handle - #4/ Use With: Scalpel Blades/ Alloy / Material: Stainless Steel. Ideal for large dissections.
Catalog No. NC9079870 Supplier Fine Science Tools Supplier No. 1000413
Product: Scalpel Handle - #4/ Use With: Scalpel Blades/ Alloy / Material: Stainless Steel. Ideal for large dissections.