Description
- Construction
- Spring-loaded, retractable design for enhanced safety
- 3 different blade shapes
- Quantity
- 10 per package
- Performance
- Great for scraping blood, paint, or other evidence from various surfaces
- Can be cleaned with 10% bleach solution or alcohol
Specifications
Specifications
| Product Type | Scalpel |
| Size | # 11 |
| Disposable | Limited-use |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Graduated | Yes |