Learn More
Med Vet International Dynarex Medicut Safety Scalpels, Sterile, #11, 10/Box
Catalog No. 502830538
Encompass_Preferred
Satisfies regulatory guidelines for safe handling of sharp edges. Stainless Steel Blade Blade retracts with one simple motion and can be permanently locked inside the handle for safe disposal Packaged in individual pouches
Catalog No. 50-283-0538 Supplier Med Vet International Supplier No. 4161
Satisfies regulatory guidelines for safe handling of sharp edges. Stainless Steel Blade Blade retracts with one simple motion and can be permanently locked inside the handle for safe disposal Packaged in individual pouches